5-Chloro-2-methoxybenzenesulphonyl chloride 97%
Supplier: Apollo Scientific
|
Danger
|
OR30141-25G
OR30141-5G
OR30141-100G
APOSOR30141-25GEA
125
GBP
APOSOR30141-25G
APOSOR30141-5G
APOSOR30141-100G
5-Chloro-2-methoxybenzenesulphonyl chloride 97%
5-Chloro-2-methoxybenzenesulphonyl chloride
Formula:
C₇H₆Cl₂O₃S MW: 241,09 g/mol Melting Pt: 100…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052959 CAS Number: 22952-32-5 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...