Methyl-2-(bromomethyl)benzoate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 2-bromomethylbenzoate
OR301372-10G
OR301372-25G
APOSOR301372-10GEA
90.5
GBP
APOSOR301372-10G
APOSOR301372-25G
Methyl-2-(bromomethyl)benzoate
Methyl 2-(bromomethyl)benzoate
Formula:
C₉H₉BrO₂ MW: 229,0706 g/mol Melting Pt: 32…33 °C Storage Temperature: Ambient |
MDL Number:
MFCD03425900 CAS Number: 2417-73-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...