3,3-Bis(bromomethyl)oxetane

Supplier: Apollo Scientific
Warning

Synonyms: 2,2-Bis(bromomethyl)-1,3-epoxypropane

OR301352-1G OR301352-5G
APOSOR301352-1GEA 44.6 GBP
APOSOR301352-1G APOSOR301352-5G
3,3-Bis(bromomethyl)oxetane
3,3-Bis(bromomethyl)oxetane
Formula: C₅H₈Br₂O
MW: 243,93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00269694
CAS Number: 2402-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR