6-Chloro-1H-pyrrolo[2,3-b]pyridine

Supplier: Apollo Scientific
Danger

OR301275-25G OR301275-5G
APOSOR301275-25GEA 202 GBP
APOSOR301275-25G APOSOR301275-5G
6-Chloro-1H-pyrrolo[2,3-b]pyridine
6-Chloro-1H-pyrrolo[2,3-b]pyridine
Formula: C₇H₅ClN₂
Storage Temperature: Ambient
MDL Number: MFCD06659666
CAS Number: 55052-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR