5-Bromo-4-chloro-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Apollo Scientific
Warning

OR301270-1G OR301270-5G
APOSOR301270-1GEA 156 GBP
APOSOR301270-1G APOSOR301270-5G
5-Bromo-4-chloro-1H-pyrrolo[2,3-b]pyridine 95%
5-Bromo-4-chloro-1H-pyrrolo[2,3-b]pyridine
Formula: C₇H₄BrClN₂
MW: 231,4771 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11046240
CAS Number: 876343-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR