6-Chloropyrazin-2-ol 98%
Supplier: Apollo Scientific
|
Warning
|
OR301254-1G
OR301254-5G
APOSOR301254-1GEA
64.3
GBP
APOSOR301254-1G
APOSOR301254-5G
6-Chloropyrazin-2-ol 98%
6-Chloropyrazin-2-ol
Formula:
C₄H₃ClN₂O MW: 130,5300 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00204104 CAS Number: 4925-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...