2-Amino-4-bromothiazole 98%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Bromo-1,3-thiazol-2-amine

OR301249-1G OR301249-5G
APOSOR301249-1GEA 77.4 GBP
APOSOR301249-1G APOSOR301249-5G
2-Amino-4-bromothiazole 98%
2-Amino-4-bromothiazole
Formula: C₃H₃BrN₂S
MW: 179,0383 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06804547
CAS Number: 502145-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR