1-Amino-2-methylpropan-2-ol 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1-Amino-2-methyl-2-propanol, 1,1-Dimethylethanolamine
OR301207-25G
OR301207-5G
APOSOR301207-25GEA
199
GBP
APOSOR301207-25G
APOSOR301207-5G
1-Amino-2-methylpropan-2-ol 98%
1-Amino-2-methylpropan-2-ol
Formula:
C₄H₁₁NO MW: 89,1362 g/mol Melting Pt: 151 °C Density: 0,94 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD01697283 CAS Number: 2854-16-2 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...