3-(Benzyloxy)cyclobutanone 97%
Supplier: Apollo Scientific
|
Warning
|
OR301159-10G
OR301159-5G
APOSOR301159-10GEA
247
GBP
APOSOR301159-10G
APOSOR301159-5G
3-(Benzyloxy)cyclobutanone 97%
3-(Benzyloxy)cyclobutanone
Formula:
C11H12O2 MW: 176,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09755991 CAS Number: 30830-27-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...