Ethyl-4-bromo-1H-indole-2-carboxylate 97%

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 4-bromo-1H-indole-2-carboxylate

OR301134-1G OR301134-250MG
APOSOR301134-1GEA 120 GBP
APOSOR301134-1G APOSOR301134-250MG
Ethyl-4-bromo-1H-indole-2-carboxylate 97%
Ethyl-4-bromo-1H-indole-2-carboxylate
Formula: C₁₁H₁₀BrNO₂
MW: 268,1066 g/mol
Melting Pt: 394…395 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12174997
CAS Number: 103858-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR