1-tert-Butyloxycarbonylpiperazine-4-acetonitrile 96%

Supplier: Apollo Scientific

Synonyms: tert-Butyl 4-(cyanomethyl)piperazine-1-carboxylate

OR300770-1G OR300770-5G
APOSOR300770-1GEA 61.7 GBP
APOSOR300770-1G APOSOR300770-5G
1-tert-Butyloxycarbonylpiperazine-4-acetonitrile 96%
1-tert-Butyloxycarbonylpiperazine-4-acetonitrile
Formula: C₁₁H₁₉N₃O₂
MW: 225,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06795916
CAS Number: 77290-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR