Quinoxaline-2-carboxylic acid methyl ester 98%
Supplier: Apollo Scientific
Synonyms:
Methyl quinoxaline-2-carboxylate
OR300583-1G
OR300583-5G
APOSOR300583-1GEA
0
GBP
APOSOR300583-1G
APOSOR300583-5G
Quinoxaline-2-carboxylic acid methyl ester 98%
Quinoxaline-2-carboxylic acid methyl ester
Formula:
C₁₀H₈N₂O₂ MW: 188,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02167640 CAS Number: 1865-11-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...