4-(Hydroxymethyl)pyrrolidin-2-one
Supplier: Apollo Scientific
|
Warning
|
OR300508-1G
OR300508-5G
APOSOR300508-1GEA
125
GBP
APOSOR300508-1G
APOSOR300508-5G
4-(Hydroxymethyl)pyrrolidin-2-one
4-(Hydroxymethyl)pyrrolidin-2-one
Formula:
C₅H₉NO₂ Melting Pt: 92…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD11845728 CAS Number: 64320-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...