4,6-Dichloro-2-(methylthio)pyrimidine-5-carbonitrile

Supplier: Apollo Scientific

OR29981-1G OR29981-5G
APOSOR29981-1GEA 64.3 GBP
APOSOR29981-1G APOSOR29981-5G
4,6-Dichloro-2-(methylthio)pyrimidine-5-carbonitrile
4,6-Dichloro-2-(methylthio)pyrimidine-5-carbonitrile
Formula: C₆H₃Cl₂N₃S
MW: 220,0800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00474490
CAS Number: 33097-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR