2-(2-Chlorophenoxy)acetamidine hydrochloride
Supplier: Apollo Scientific
|
Warning
|
OR29776-100MG
OR29776-500MG
APOSOR29776-100MGEA
13.2
GBP
APOSOR29776-100MG
APOSOR29776-500MG
2-(2-Chlorophenoxy)acetamidine hydrochloride
2-(2-Chlorophenoxy)acetamidine hydrochloride
Formula:
C₈H₁₀Cl₂N₂O MW: 221,09 g/mol Melting Pt: 165…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173805 CAS Number: 58403-03-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...