1-Chloro-5-iodopentane

Supplier: Apollo Scientific
Warning

OR2936-25G OR2936-5G
APOSOR2936-25GEA 37.5 GBP
APOSOR2936-25G APOSOR2936-5G
1-Chloro-5-iodopentane
1-Chloro-5-iodopentane
Formula: C₅H₁₀ClI
Boiling Pt: 75…77 °C/3mmHg
Density: 1,663 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039416
CAS Number: 60274-60-4
EINECS: 262-137-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR