3-(3-Methyl-1H-indol-1-yl)propanoic acid

Supplier: Apollo Scientific
Warning

OR29133-1G OR29133-5G
APOSOR29133-1GEA 122 GBP
APOSOR29133-1G APOSOR29133-5G
3-(3-Methyl-1H-indol-1-yl)propanoic acid
3-(3-Methyl-1H-indol-1-yl)propanoic acid
Formula: C₁₂H₁₃NO₂
Melting Pt: 84…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052088
CAS Number: 57662-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR