3,4-Thiophenedicarboxylic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Thiophene-3,4-dicarboxylic acid
OR2907-25G
OR2907-5G
APOSOR2907-25GEA
173
GBP
APOSOR2907-25G
APOSOR2907-5G
3,4-Thiophenedicarboxylic acid
3,4-Thiophenedicarboxylic acid
Formula:
C₆H₄O₄S MW: 172,15 g/mol Melting Pt: 227…231 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00067079 CAS Number: 4282-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...