(4-Hydroxyphenyl)acetamide
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Hydroxyphenylacetamide
OR28890-100G
OR28890-250G
APOSOR28890-100GEA
13.2
GBP
APOSOR28890-100G
APOSOR28890-250G
(4-Hydroxyphenyl)acetamide
(4-Hydroxyphenyl)acetamide
Formula:
C₈H₉NO₂ MW: 151,16 g/mol Melting Pt: 173…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017145 CAS Number: 17194-82-0 EINECS: 241-235-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...