2,4-Diphenyl-1H-imidazole 98%
Supplier: Apollo Scientific
Synonyms:
2,5-Diphenyl-1H-imidazole
OR2884-10G
OR2884-5G
APOSOR2884-10GEA
397
GBP
APOSOR2884-10G
APOSOR2884-5G
2,4-Diphenyl-1H-imidazole 98%
2,4-Diphenyl-1H-imidazole
Formula:
C₁₅H₁₂N₂ MW: 220,28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07772899 CAS Number: 670-83-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...