4-Bromo-6-chloro-o-cresol 97%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Bromo-2-chloro-6-methylphenol

OR28761-10G OR28761-50G
APOSOR28761-10GEA 111 GBP
APOSOR28761-10G APOSOR28761-50G
4-Bromo-6-chloro-o-cresol 97%
4-Bromo-6-chloro-o-cresol
Formula: C₇H₆BrClO
MW: 221,4800 g/mol
Melting Pt: 45…47 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002167
CAS Number: 7530-27-0
EINECS: 231-394-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR