2-Amino-4-methylpyridine
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Aminopicoline, 2-Amino-4-picoline, 4-Methyl-2-pyridylamine, 4-Methylpyridin-2-amine
OR28757-25G
OR28757-100G
OR28757-500G
APOSOR28757-25GEA
39.4
GBP
APOSOR28757-25G
APOSOR28757-100G
APOSOR28757-500G
2-Amino-4-methylpyridine
2-Amino-4-methylpyridine
Formula:
C₆H₈N₂ MW: 108,14 g/mol Boiling Pt: 230 °C Melting Pt: 96…101 °C Flash Pt: 118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006321 CAS Number: 695-34-1 EINECS: 211-780-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...