2-Acetylfuran

Supplier: Apollo Scientific
Danger

Synonyms: 2-Furyl methyl ketone, 1-(Furan-2-yl)ethanone

OR28745-100G OR28745-250G OR28745-50G
APOSOR28745-100GEA 72.2 GBP
APOSOR28745-100G APOSOR28745-250G APOSOR28745-50G
2-Acetylfuran
2-Acetylfuran
Formula: C₆H₆O₂
MW: 110,11 g/mol
Boiling Pt: 171…173 °C
Melting Pt: 26…30 °C
Density: 1,098 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003242
CAS Number: 1192-62-7
EINECS: 214-757-1
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR