4,5-Dimethoxyanthranilic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Amino-4,5-dimethoxybenzoic acid, 6-Aminoveratric acid
OR28629-100G
OR28629-25G
APOSOR28629-100GEA
91.8
GBP
APOSOR28629-100G
APOSOR28629-25G
4,5-Dimethoxyanthranilic acid 98%
4,5-Dimethoxyanthranilic acid
Formula:
C₉H₁₁NO₄ MW: 197,19 g/mol Melting Pt: 169…173 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011671 CAS Number: 5653-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...