Pivalaldehyde (trimethylacetaldehyde) 95%

Supplier: Apollo Scientific
Danger

Synonyms: Pivalaldehyde, Pivaldehyde, Trimethylacetaldehyde, 2,2-Dimethylpropionaldehyde, 2,2-Dimethylpropanal

OR28610-25G OR28610-10G OR28610-100G
APOSOR28610-25GEA 62.9 GBP
APOSOR28610-25G APOSOR28610-10G APOSOR28610-100G
Pivalaldehyde (trimethylacetaldehyde) 95%
Pivalaldehyde (Trimethylacetaldehyde)
Formula: C₅H₁₀O
MW: 86,13 g/mol
Boiling Pt: 71…74 °C
Melting Pt: 6 °C
Density: 0,783 g/cm³ (25 °C)
Flash Pt: –16 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006962
CAS Number: 630-19-3
UN: 1989
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR