3-Chloromethylbenzamide

Supplier: Apollo Scientific
Danger

Synonyms: 3-(Chloromethyl)benzamide

OR28458-1G OR28458-5G
APOSOR28458-1GEA 0 GBP
APOSOR28458-1G APOSOR28458-5G
3-Chloromethylbenzamide
3-Chloromethylbenzamide
Formula: C₈H₈ClNO
MW: 169,61 g/mol
Melting Pt: 125…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00175948
CAS Number: 135654-16-9
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR