5-Isoquinolinol 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Hydroxyisoquinoline
OR2817-1G
OR2817-5G
APOSOR2817-1GEA
39.3
GBP
APOSOR2817-1G
APOSOR2817-5G
5-Isoquinolinol 97%
5-Isoquinolinol
Formula:
C₉H₇NO Melting Pt: 213…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006906 CAS Number: 2439-04-5 EINECS: 219-456-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...