2,4-Dichloropyrimidine-5-carbonyl chloride 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2,4-Dichloro-5-pyrimidinecarbonyl chloride
OR2807-1G
OR2807-250MG
OR2807-5G
APOSOR2807-1GEA
151
GBP
APOSOR2807-1G
APOSOR2807-250MG
APOSOR2807-5G
2,4-Dichloropyrimidine-5-carbonyl chloride 97%
2,4-Dichloropyrimidine-5-carbonyl chloride
Formula:
C₅HCl₃N₂O MW: 211,44 g/mol Boiling Pt: 67…70 °C/0,1mmHg Density: 1,645 g/cm³ (25 °C) Flash Pt: 93,3 °C Storage Temperature: Freezer |
MDL Number:
MFCD09742432 CAS Number: 2972-52-3 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...