3-Chloroisonicotinonitrile

Supplier: Apollo Scientific
Warning

OR2798-1G OR2798-250MG
APOSOR2798-1GEA 18.5 GBP
APOSOR2798-1G APOSOR2798-250MG
3-Chloroisonicotinonitrile
3-Chloroisonicotinonitrile
Formula: C₆H₃ClN₂
MW: 138,56 g/mol
Melting Pt: 71…74 °C
Storage Temperature: Ambient
MDL Number: MFCD05663706
CAS Number: 68325-15-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR