1H-Pyrazolo[3,4-d]pyrimidin-4-amine 95%

Supplier: Apollo Scientific
Danger

Synonyms: 4-Amino-1H-pyrazolo[3,4-d]pyrimidine

OR27867-25G OR27867-100G OR27867-500G
APOSOR27867-25GEA 118 GBP
APOSOR27867-25G APOSOR27867-100G APOSOR27867-500G
1H-Pyrazolo[3,4-d]pyrimidin-4-amine 95%
1H-Pyrazolo[3,4-d]pyrimidin-4-amine
Formula: C₅H₅N₅
MW: 135,13 g/mol
Melting Pt: >325 °C
Storage Temperature: Ambient
MDL Number: MFCD00005688
CAS Number: 2380-63-4
EINECS: 219-174-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR