2-Methoxypyrimidine-5-carbaldehyde

Supplier: Apollo Scientific
Warning

OR2778-100MG OR2778-1G
APOSOR2778-100MGEA 42 GBP
APOSOR2778-100MG APOSOR2778-1G
2-Methoxypyrimidine-5-carbaldehyde
2-Methoxypyrimidine-5-carbaldehyde
Formula: C₆H₆N₂O₂
MW: 138,1200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03446732
CAS Number: 90905-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR