8-Methoxy-2-oxo-2H-chromene-3-carboxylic acid

Supplier: Apollo Scientific

OR27771-1G OR27771-5G
APOSOR27771-1GEA 98.3 GBP
APOSOR27771-1G APOSOR27771-5G
8-Methoxy-2-oxo-2H-chromene-3-carboxylic acid
8-Methoxy-2-oxo-2H-chromene-3-carboxylic acid
Formula: C₁₁H₈O₅
Storage Temperature: Ambient
MDL Number: MFCD01116499
CAS Number: 2555-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR