N,N-Dimethylpyridin-3-amine 98%
Supplier: Apollo Scientific
OR27549-5G
OR27549-25G
APOSOR27549-5GEA
231
GBP
APOSOR27549-5G
APOSOR27549-25G
N,N-Dimethylpyridin-3-amine 98%
N,N-Dimethylpyridin-3-amine
Formula:
C₇H₁₀N₂ MW: 122,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00234349 CAS Number: 18437-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...