6-Chloronicotinoyl chloride

Supplier: Apollo Scientific
Danger

OR27408-100G OR27408-25G
APOSOR27408-100GEA 464 GBP
APOSOR27408-100G APOSOR27408-25G
6-Chloronicotinoyl chloride
6-Chloronicotinoyl chloride
Formula: C₆H₃Cl₂NO
Boiling Pt: 84…85 °C/3mmHg
Melting Pt: 48…52 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00051775
CAS Number: 58757-38-3
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR