4-(1-Adamantyl)-6-(methylthio)-1,3,5-trazin-2-amine
Supplier: Apollo Scientific
OR27323-10G
OR27323-1G
OR27323-5G
APOSOR27323-10GEA
915
GBP
APOSOR27323-10G
APOSOR27323-1G
APOSOR27323-5G
4-(1-Adamantyl)-6-(methylthio)-1,3,5-trazin-2-amine
4-(1-Adamantyl)-6-(methylthio)-1,3,5-trazin-2-amine
Formula:
C₁₄H₂₀N₄S Storage Temperature: Ambient |
MDL Number:
MFCD00078505 CAS Number: 175204-72-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...