Ethyl-2-amino-3-hydroxypropanoate hydrochloride 98%

Supplier: Apollo Scientific
Warning

Synonyms: H-DL-Ser-OEt·HCl, DL-serine ethyl ester hydrochloride

OR26790-1G OR26790-5G OR26790-25G
APOSOR26790-1GEA 13.2 GBP
APOSOR26790-1G APOSOR26790-5G APOSOR26790-25G
Ethyl-2-amino-3-hydroxypropanoate hydrochloride 98%
Ethyl-2-amino-3-hydroxypropanoate hydrochloride
Formula: C₅H₁₁NO₃·HCl
MW: 169,61 g/mol
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00067544
CAS Number: 3940-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR