L-tert-Leucinol 98%

Supplier: Apollo Scientific
Warning

Synonyms: (S)-tert-Leucinol, (S)-2-Amino-3,3-dimethylbutan-1-ol, (S)-2-Amino-3,3-dimethylbutanol

OR26748-10G OR26748-5G
APOSOR26748-10GEA 153 GBP
APOSOR26748-10G APOSOR26748-5G
L-tert-Leucinol 98%
L-tert-Leucinol
Formula: C₆H₁₅NO
MW: 117,19 g/mol
Boiling Pt: 70 °C/0,5mmHg
Melting Pt: 30…34 °C
Density: 0,9 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00192250
CAS Number: 112245-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR