6-Methyluracil

Supplier: Apollo Scientific

Synonyms: 6-Methylpyrimidine-2,4(1H,3H)-dione, 4-Methyluracil, 6-Methyl-2,4(1H,3H)-pyrimidinedione

OR26591-1KG OR26591-500G
APOSOR26591-1KGEA 127 GBP
APOSOR26591-1KG APOSOR26591-500G
6-Methyluracil
6-Methyluracil
Formula: C₅H₆N₂O₂
MW: 126,11 g/mol
Melting Pt: 318 °C
Storage Temperature: Ambient
MDL Number: MFCD00006028
CAS Number: 626-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR