2-[(4-Nitrophenoxy)methyl]oxirane

Supplier: Apollo Scientific
Warning

OR26560-1G OR26560-250MG OR26560-5G
APOSOR26560-1GEA 132 GBP
APOSOR26560-1G APOSOR26560-250MG APOSOR26560-5G
2-[(4-Nitrophenoxy)methyl]oxirane
2-[(4-Nitrophenoxy)methyl]oxirane
Formula: C₉H₉NO₄
Melting Pt: 62…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00041845
CAS Number: 5255-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR