5-Methylbenzo[b]thiophene
Supplier: Apollo Scientific
|
Warning
|
OR26010-1G
OR26010-250MG
APOSOR26010-1GEA
34.2
GBP
APOSOR26010-1G
APOSOR26010-250MG
5-Methylbenzo[b]thiophene
5-Methylbenzo[b]thiophene
Formula:
C₉H₈S Boiling Pt: 71…73 °C/0,6mmHg Melting Pt: 32…37 °C Density: 1,11 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00052509 CAS Number: 14315-14-1 EINECS: 238-256-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...