3-Methoxy-4-(methoxycarbonyl)thiophene-2-sulphonyl chloride 97%

Supplier: Apollo Scientific

OR25576-1G OR25576-5G
APOSOR25576-1GEA 189 GBP
APOSOR25576-1G APOSOR25576-5G
3-Methoxy-4-(methoxycarbonyl)thiophene-2-sulphonyl chloride 97%
3-Methoxy-4-(methoxycarbonyl)thiophene-2-sulphonyl chloride
Formula: C₇H₇ClO₅S₂
Melting Pt: 94…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00173848
CAS Number: 175203-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR