Benzofurazan

Supplier: Apollo Scientific
Warning

Synonyms: 2,1,3-Benzoxadiazole, Benzo[c][1,2,5]oxadiazole

OR25510-1G OR25510-25G OR25510-5G
APOSOR25510-1GEA 0 GBP
APOSOR25510-1G APOSOR25510-25G APOSOR25510-5G
Benzofurazan
Benzofurazan
Formula: C₆H₄N₂O
MW: 120,11 g/mol
Boiling Pt: 75…85 °C/20mmHg
Melting Pt: 47…55 °C
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00068064
CAS Number: 273-09-6
EINECS: 608-080-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR