Phenyl chloroacetate

Supplier: Apollo Scientific
Danger

OR25299-1G OR25299-250MG OR25299-5G OR25299-2.5G
APOSOR25299-1GEA 249 GBP
APOSOR25299-1G APOSOR25299-250MG APOSOR25299-5G APOSOR25299-2.5G
Phenyl chloroacetate
Phenyl chloroacetate
Formula: C₈H₇ClO₂
Storage Temperature: Ambient
CAS Number: 620-73-5
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR