1-(3-Amino-4-chlorophenyl)ethanone

Supplier: Apollo Scientific
Warning

Synonyms: 1-(3-Amino-4-chlorophenyl)ethan-1-one, 3'-Amino-4'-chloroacetophenone

OR24995-1G OR24995-5G
APOSOR24995-1GEA 21.1 GBP
APOSOR24995-1G APOSOR24995-5G
1-(3-Amino-4-chlorophenyl)ethanone
1-(3-Amino-4-chlorophenyl)ethanone
Formula: C₈H₈ClNO
MW: 169,61 g/mol
Melting Pt: 104…109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00122619
CAS Number: 79406-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR