Methyl-3,5-dihydroxybenzoate

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 3,5-dihydroxybenzoate, 3,5-Dihydroxybenzoic acid methyl ester

OR24884-100G OR24884-25G OR24884-500G
APOSOR24884-100GEA 27.6 GBP
APOSOR24884-100G APOSOR24884-25G APOSOR24884-500G
Methyl-3,5-dihydroxybenzoate
Methyl 3,5-dihydroxybenzoate
Formula: C₈H₈O₄
MW: 168,15 g/mol
Melting Pt: 164…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00002289
CAS Number: 2150-44-9
EINECS: 218-426-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR