2,5-Dimethyl-1,3,4-thiadiazole

Supplier: Apollo Scientific
Warning

OR24865-25G OR24865-5G
APOSOR24865-25GEA 202 GBP
APOSOR24865-25G APOSOR24865-5G
2,5-Dimethyl-1,3,4-thiadiazole
2,5-Dimethyl-1,3,4-thiadiazole
Formula: C₄H₆N₂S
MW: 114,17 g/mol
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00020722
CAS Number: 27464-82-0
EINECS: 248-473-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR