1-Bromo-3-ethyladamantane 95+%

Supplier: Apollo Scientific
Warning

OR2484-1G OR2484-5G
APOSOR2484-1GEA 78.5 GBP
APOSOR2484-1G APOSOR2484-5G
1-Bromo-3-ethyladamantane 95+%
1-Bromo-3-ethyladamantane
Formula: C₁₂H₁₉Br
Boiling Pt: 124…125 °C/7mmHg
Melting Pt: 23…24 °C
Storage Temperature: Ambient
MDL Number: MFCD00274777
CAS Number: 878-61-5
EINECS: 227-553-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR