3-(2-Bromoethyl)-1H-indole 97%

Supplier: Apollo Scientific
Warning

Synonyms: 3-(2-Bromoethyl)indole

OR24580-1G OR24580-5G
APOSOR24580-1GEA 74.8 GBP
APOSOR24580-1G APOSOR24580-5G
3-(2-Bromoethyl)-1H-indole 97%
3-(2-Bromoethyl)-1H-indole
Formula: C₁₀H₁₀BrN
MW: 224,101 g/mol
Melting Pt: 97…99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00130155
CAS Number: 3389-21-7
EINECS: 222-219-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR