Bromobenzene 99%

Supplier: Apollo Scientific
Warning

Synonyms: Phenylbromide, Phenyl bromide

OR2446-2.5KG OR2446-500G
APOSOR2446-2.5KGEA 165 GBP
APOSOR2446-2.5KG APOSOR2446-500G
Bromobenzene 99%
Bromobenzene
Formula: C₆H₅Br
MW: 157,01 g/mol
Boiling Pt: 155…156 °C
Melting Pt: –31 °C
Density: 1,491 g/cm³ (25 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00000055
CAS Number: 108-86-1
EINECS: 203-623-8
UN: 2514
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR