5-Amino-4-bromo-1-methyl-1H-pyrazole

Supplier: Apollo Scientific
Warning

Synonyms: 4-Bromo-1-methyl-1H-pyrazol-5-amine

OR24415-1G OR24415-5G
APOSOR24415-1GEA 42 GBP
APOSOR24415-1G APOSOR24415-5G
5-Amino-4-bromo-1-methyl-1H-pyrazole
5-Amino-4-bromo-1-methyl-1H-pyrazole
Formula: C₄H₆BrN₃
MW: 176,02 g/mol
Melting Pt: 96…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00068157
CAS Number: 105675-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR